missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Antibody (661002) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB63341JF525
This item is not returnable.
View return policy
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot
Specifications
| LAT | |
| Monoclonal | |
| Janelia Fluor 525 | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| E. coli-derived recombinant human LAT, His30-Gln113, Accession # O43561 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| 661002 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 27040 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction