missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Antibody (LAT.10-17), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00289B
This item is not returnable.
View return policy
Description
LAT Monoclonal specifically detects LAT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| LAT | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| LAT | |
| The immunogen for this antibody is LAT. | |
| 100 μL | |
| Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
| 27040 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| LAT.10-17 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction