missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latent TGF-beta bp1 Antibody (MM0447-5L38), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11783IR
This item is not returnable.
View return policy
Description
Latent TGF-beta bp1 Monoclonal specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Latent TGF-beta bp1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LTBP1 | |
| Human recombinant Latent TGF-beta bp1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0447-5L38 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1 | |
| Mouse | |
| Protein G purified | |
| Ovarian Carcinoma Cell Markers | |
| 4052 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction