missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latent TGF-beta bp2/LTBP-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88411-25ul
This item is not returnable.
View return policy
Description
Latent TGF-beta bp2/LTBP-2 Polyclonal specifically detects Latent TGF-beta bp2/LTBP-2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Latent TGF-beta bp2/LTBP-2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| C14orf141, GLC3D, latent transforming growth factor beta binding protein 2, latent-transforming growth factor beta-binding protein 2, LTBP-2, LTBP3chromosome 14 open reading frame 141, MSTP031 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| LTBP2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ERSPNLRRSSAAGEGTLARAQPPAPQSPPAPQSPPAGTLSGLSQTHPSQQHVGLSRTVRLHPTATASSQLSSNALPPGPGLEQRDGTQQAVPLEHPSSPWGLNLTEKIKKIKIVFTPTICKQTCARGHCANSCERG | |
| 25 μL | |
| Cell Cycle and Replication | |
| 4053 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction