missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latexin Antibody (009), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90379H
This item is not returnable.
View return policy
Description
Latexin Monoclonal specifically detects Latexin in Mouse samples. It is validated for ELISA.
Specifications
| Latexin | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Human LXN | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 9 | |
| ELISA | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Latexin (NP_058033.2; Glu 2-Glu 222). | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 56925 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction