missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latexin Antibody (OTI1E10), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72069G
This item is not returnable.
View return policy
Description
Latexin Monoclonal specifically detects Latexin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| Latexin | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LXN | |
| Full length recombinant protein of human Latexin (NP_064554) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1E10 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| Mouse | |
| Affinity Purified | |
| Immunology, Innate Immunity | |
| 56925.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction