missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latexin Antibody (OTI1E10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72069MFV500
This item is not returnable.
View return policy
Description
Latexin Monoclonal antibody specifically detects Latexin in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| Latexin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| OTI1E10 | |
| Flow Cytometry | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| Full length human recombinant protein of human LXN(NP_064554) produced in HEK293T cell. | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 56925 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction