Learn More
Hach Lange™ Laton Total Nitrogen Cuvette Tests
Product Code. 10329662
Change view
Click to view available options
Range:
1 to 16mg/L TNb
20 to 100mg/L TNb
5 to 40mg/L TNb
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10329662 5 to 40mg/L TNb Each
10409464 20 to 100mg/L TNb Each
10742357 1 to 16mg/L TNb Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10329662 Supplier Hach Lange™ Supplier No. LCK238
Specifications
Certifications/Compliance EN ISO 11905-1
No. of Tests 25
Range 5 to 40mg/L TNb
Storage Requirements 15°C to 25°C
Test Method Koroleff Digestion (Peroxodisulphate), and Photometric Detection with 2.6-Dimethylphenol
Type Laton Total Nitrogen Cuvette Test
compliance-icons compliance-icons compliance-icons
Product Identifier
  • LatoN, Sample cuvette
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric acid, Phosphoric acid, Sodiumhydroxide, Potassiumnitrate, dipotassium peroxodisulphate,