Learn More
LATS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17049-1-AP-20UL
This item is not returnable.
View return policy
Description
Plays a role in maintece of the normal epithelial architecture through the repression of SNAI1 transcription in a histone deacetylase-dependent manner, and thus the regulation of E-cadherin levels.Specifications
| LATS1 | |
| Polyclonal | |
| Unconjugated | |
| LATS1 | |
| h warts, LATS1, WARTS, WARTS protein kinase, wts | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16798, 308265, 9113 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95835, Q8BYR2 | |
| LATS1 | |
| LATS1 Fusion Protein Ag10709 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |