Learn More
Lauroyl-L-carnitine-d3 Hydrochloride, TRC
Product Code. 30478472
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478472 2.5 mg 2.5mg
30474909 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478472 Supplier Toronto Research Chemicals Supplier No. TRCL1786822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2687960-76-3
Molecular Formula C19 D3 H35 N O4 . Cl
Molecular Weight (g/mol) 382.9808
Synonym 1-Propanaminium, 3-carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxododecyl)oxy]-, chloride (1:1), (2R)- (ACI), Dodecanoyl-L-carnitine-d3 HCl (N-methyl-d3), Lauroyl-L-carnitine-d3 Chloride, Lauroyl-L-carnitine-D3 (N-methyl-D3) Chloride, (2R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[(1-oxododecyl)oxy]-1-propanaminium chloride
IUPAC Name [(2R)-3-carboxy-2-dodecanoyloxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
SMILES [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCCCCCC
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C19H37NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-19(23)24-17(15-18(21)22)16-20(2,3)4;/h17H,5-16H2,1-4H3;1H/t17-;/m1./s1/i2D3;
Formula Weight 382.2678
Chemical Name or Material Lauroyl-L-carnitine-d3 Hydrochloride