missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Layilin Antibody (328024), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2637E
This item is not returnable.
View return policy
Description
Layilin Monoclonal antibody specifically detects Layilin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Layilin | |
| Monoclonal | |
| DyLight 405 | |
| FLJ30977, FLJ31092, layilin | |
| Mouse myeloma cell line NS0-derived recombinant human Layilin, Ala22-Glu220, Accession # NP_849156 | |
| 0.1 mL | |
| Signal Transduction | |
| 143903 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| 328024 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction