missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Layilin Antibody (328024) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2637JF585
This item is not returnable.
View return policy
Description
Layilin Monoclonal antibody specifically detects Layilin in Human,Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,CyTOF
Specifications
| Layilin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FLJ30977, FLJ31092, layilin | |
| Mouse myeloma cell line NS0-derived recombinant human Layilin, Ala22-Glu220, Accession # NP_849156 | |
| 0.1 mL | |
| Signal Transduction | |
| 143903 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Western Blot, CyTOF | |
| 328024 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction