missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Layilin Antibody (MM0448-4F33), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11767MFV500
This item is not returnable.
View return policy
Description
Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for Western Blot
Specifications
| Layilin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0448-4F33 | |
| Western Blot | |
| FLJ30977, FLJ31092, layilin | |
| Human recombinant Layilin | |
| 0.1 mL | |
| Signal Transduction | |
| 143903 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction