missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Layilin Antibody (OTI4C11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72402CL1
This item is not returnable.
View return policy
Description
Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| Layilin | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ30977, FLJ31092, layilin | |
| Full length human recombinant protein of human LAYN(NP_849156) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 143903 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| OTI4C11 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu