missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Layilin Antibody (OTI4D3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72403PE
This item is not returnable.
View return policy
Description
Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for Western Blot
Specifications
| Layilin | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI4D3 | |
| Western Blot | |
| FLJ30977, FLJ31092, layilin | |
| Full length human recombinant protein of human LAYN(NP_849156) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 143903 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction