missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A Antibody (2312D), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-75924CL1
This item is not returnable.
View return policy
Description
LC3A Monoclonal antibody specifically detects LC3A in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| LC3A | |
| Monoclonal | |
| CoraFluor 1 | |
| ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, MAP1 light chain 3-like protein 1, MAP1A/1B light chain 3 A, MAP1A/MAP1B LC3 A, MAP1A/MAP1B light chain 3 A, MAP1ALC3, MAP1BLC3, MAP1LC3A, Microtubule-associated protein 1 light chain 3 alpha, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A | |
| This LC3A Antibody (2312D) was immunized with a partial synthetic peptide from the C-terminal of human LC3A [UniProt Q9H492] | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Macroautophagy, Membrane Trafficking and Chaperones, Neuroscience, Signal Transduction | |
| 84557 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 2312D | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction