missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3A Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19167PE
This item is not returnable.
View return policy
Description
LC3A Polyclonal specifically detects LC3A in Human, Mouse, Rat, Bovine, Zebrafish samples. It is validated for Flow Cytometry.
Specifications
| LC3A | |
| Polyclonal | |
| Flow Cytometry | |
| Q9H492 | |
| MAP1LC3A | |
| Genomic sequence made to an N-terminal portion of the human LC3A protein [Swiss-Prot# Q9H492]. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Although specificity between LC3A and LC3B has not been tested, this antibody was created to a peptide that has 100% identity to LC3A and 62% identity to LC3B. | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A | |
| Rabbit | |
| 14 kDa | |
| 0.1 mL | |
| Autophagy, Cancer, Cellular Markers, Cytoskeleton Markers, Hypoxia, Phospho Specific, Signal Transduction | |
| 84557 | |
| Human, Mouse, Rat, Bovine, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction