missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LC3A Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μl
Unit Size:
100µL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human MAP1LC3A. The LC3A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 84557 |
| Species | Human |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Antibody Competition |
| Gene Symbol | MAP1LC3A |
| Molecular Weight (g/mol) | 25 kDa |
| Quantity | 100 μl |
| Regulatory Status | RUO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction