missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2220AF750
This item is not returnable.
View return policy
Description
LC3B Polyclonal antibody specifically detects LC3B in Human, Mouse, Rat, Porcine, Alligator, Avian, Bacteria, Bovine, Canine, Chicken, Chinese Hamster, Guinea Pig, Hamster, Invertebrate, Monkey, Primate, Rabbit, Golden Syrian Hamster, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Knockdown.
Specifications
| LC3B | |
| Polyclonal | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockout Validated, KnockDown Validated | |
| ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Rabbit | |
| 14.688 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Pig, Alligator, Avian, Bacteria, Bovine, Canine, Chicken, Guinea Pig, Hamster, Invertebrate, Monkey, Primate, Rabbit, Golden Syrian Hamster, Zebrafish | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MAP1LC3B | |
| Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8] | |
| Affinity Purified | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience | |
| 81631.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction