missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2220CL1
This item is not returnable.
View return policy
Description
LC3B Polyclonal antibody specifically detects LC3B in Alligator,Avian,Bacteria,Bovine,Canine,Chicken,Chinese Hamster,Golden Syrian Hamster,Guinea Pig,Hamster,Human,Invertebrate,Monkey,Mouse,Porcine,Primate,Rabbit,Rat,Zebrafish samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunoblot,Proximity Ligation Assay,SDS-Page,Simple Western,ChIP assay (ChIP),Gene Knock-Out,KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| LC3B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Alligator, Avian, Bacteria, Bovine, Canine | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B | |
| Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100). [UniProt# Q9GZQ8] | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Neuroscience | |
| 81631 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction