missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCA5 Monoclonal Antibody (OTI1A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812737
This item is not returnable.
View return policy
Description
Might be involved in minus end-directed microtubule transport.Specifications
| LCA5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86VQ0 | |
| LCA5 | |
| Full length human recombit protein of human LCA5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1A3 | |
| Unconjugated | |
| LCA5 | |
| C6orf152 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 167691 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction