missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556050
This item is not returnable.
View return policy
Description
Immunogen sequence: NSGNVRSPAS PNEFAFGSYV PSFAKTSERS NPFSQKSSFL DFQRNSMEKL SKDGVDLITR KEKKANLMEQ LFGASG Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.
Might be involved in minus end-directed microtubule transport.
Specifications
| LCA5 | |
| Polyclonal | |
| Unconjugated | |
| LCA5 | |
| 4930431B11Rik; 5730406O13Rik; AV274874; C6orf152; LCA5; LCA5, lebercilin; Leber congenital amaurosis 5; Leber congenital amaurosis 5 (human); leber congenital amaurosis 5 protein; Leber congenital amaurosis 5 protein homolog; lebercilin; ORF64; RGD1308555 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167691 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86VQ0 | |
| LCA5 | |
| Recombinant protein corresponding to Human LCA5. Recombinant protein control fragment (Product #RP-104092). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction