missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12243-1-AP-150UL
This item is not returnable.
View return policy
Description
LCAT encodes the extracellular cholesterol esterifying enzyme, lecithin-cholesterol acyltransferase. The esterification of cholesterol is required for cholesterol transport.Specifications
| LCAT | |
| Polyclonal | |
| Unconjugated | |
| LCAT | |
| LCAT | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16816, 24530, 3931 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04180, P16301, P18424 | |
| LCAT | |
| LCAT Fusion Protein Ag2887 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction