missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxoid UK LCAT Selective Supplement 10 vials
Click to view available options
:
LCAT Selective Supplement 10 vials
Description
A selective supplement for the isolation of pathogenic neisseria from CM0367 GC Agar BaseEach vial is sufficient for 2 Litres of medium
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Country of Origin | UK |
| Format | Box |
| Media Type | Supplement |
| Product Type | Selective Supplement |
| For Use With (Application) | A Selective Supplement for Use with New York City Medium (CM0367) for The Isolation of Pathogenic Neisseria. |
| Packaging Type | 10 vials |
| Quantity | 10/Pk. |
| Species | Neisseria species |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction