Learn More
Fisherbrand™ LCD Thermometer
Monitor temperature with this thermometer. Fisherbrand™ LCD Thermometers feature a probe for refrigerator and freezer use.
Brand: Fisherbrand™
Additional Details : Weight : 0.07380kg
Description
- LCD Thermometer
- Dual temp. range -50 to 70°C/-58 to 158°F
- Supplied with AAA batteries
- Temperature alert alarm
Specifications
158°F | |
-58°F | |
-58°F to +158°F |
70°C | |
-50°C | |
-50°C to +70°C |