missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach™ LCK Cuvette Tests
Click to view available options
Analytes:
Chloride (Cl)
Chromium VI and Total Cr
Dissolved Sulphide
Total Organic Carbon
Unit Size:
Each
Pack of 25
Description
Hach LCK Cuvette Tests are officially approved for legally required consent limits. With the help of Addista standard solutions and round robin test solutions, they provide the assurance that analytical results are always correct. The photometer identifies the cuvette test from its barcode (IBR+) and calculates the result immediately. Cuvettes provide precision and safety using the Dosicap Zip that eliminated contact with reagents.
- Pre-dosed, ready-to-use reagents with no rinsing steps needed
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Specifications
Specifications
| Analytes | Chromium VI and Total Cr |
| Certifications/Compliance | EN ISO 11083, DIN 38405-D24 |
| Color | Green |
| Format | Pre-dosed Tubes |
| For Use With (Application) | Water quality testing |
| For Use With (Equipment) | Photometer (DR3900, DR6000, DR1900, DR2800, DR3800, DR5000), Addista standard solutions |
| Includes | Fully equipped box, reagent set, digestion for Total Chromium |
| No. of Tests | 25 |
| Range | 0.03 to 1.0mg/L Cr VI or Cr Total |
| Storage Requirements | 2°C to 8°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction