missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCMT1 Monoclonal Antibody (OTI1G5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525598
This item is not returnable.
View return policy
Description
LCMT1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.
Specifications
| LCMT1 | |
| Monoclonal | |
| 0.62 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UIC8 | |
| LCMT1 | |
| Full length human recombinant protein of LCMT1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1G5 | |
| Unconjugated | |
| LCMT1 | |
| [phosphatase 2A protein]-leucine-carboxy methyltransferase; [Phosphatase 2A protein]-leucine-carboxy methyltransferase 1; AL033290; CGI-68; Lcmt; Lcmt1; LCMT-1; Leucine carboxyl methyltransferase 1; PPMT1; protein phosphatase methyltransferase 1; protein-leucine O-methyltransferase | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51451 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction