missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCMT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558774
This item is not returnable.
View return policy
Description
Immunogen sequence: FKSEDNNTED LKVTITKAGR KDDSTLCCWR HSTTEVSCQN QEYLFVYGGR SVVEPVLSDW HFLHVGTMAW VRIPVEGEVP EARHS Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 68%.
TYW4 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW4 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.
Specifications
| LCMT2 | |
| Polyclonal | |
| Unconjugated | |
| LCMT2 | |
| D330024M17; KIAA0547; LCMT2; Leucine carboxyl methyltransferase 2; p21WAF1/CIP1 promoter-interacting protein; PPM2; tRNA wybutosine-synthesizing protein 4; tRNA wybutosine-synthesizing protein 4 homolog; tRNA yW-synthesizing protein 4; tRNA yW-synthesizing protein 4 homolog; tRNA(Phe) (7-(3-amino-3-(methoxycarbonyl)propyl)wyosine(37)-N)-methoxycarbonyltransferase; tRNA(Phe) (7-(3-amino-3-carboxypropyl)wyosine(37)-O)-methyltransferase; tRNA-Wybutosine-synthesizing protein 4; tRNA-yW synthesizing protein 4; TYW4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9836 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60294 | |
| LCMT2 | |
| Recombinant protein corresponding to Human LCMT2. Recombinant protein control fragment (Product #RP-98132). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction