missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LCN12 Polyclonal Antibody

Product Code. 15526692
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15526692 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15526692 Supplier Invitrogen™ Supplier No. PA544898

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: GNQFQGEWFV LGLAGNSFRP EHRALLNAFT ATFELSDDGR FEVWNAMTRG Sequence homology: Human: 100%; Mouse: 79%.

LCN12 may play a role in male fertility. LCN12 may act as a retinoid carrier protein within the epididymis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LCN12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LCN12
Gene Accession No. Q6JVE5
Gene Alias 9230102M18Rik; Epididymal-specific lipocalin-12; Lcn12; lipocalcin 12; lipocalin 12
Gene Symbols LCN12
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LCN12 (aa 36-85).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286256
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.