missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDAH Antibody (OTI2G9), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72122MFV610
This item is not returnable.
View return policy
Description
LDAH Monoclonal antibody specifically detects LDAH in Human samples. It is validated for Flow Cytometry
Specifications
| LDAH | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 60526 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI2G9 | |
| Flow Cytometry | |
| chromosome 2 open reading frame 43, FLJ21820, hypothetical protein LOC60526, lipid droplet associated hydrolase | |
| Full length human recombinant protein of human C2orf43(NP_068744) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction