missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDB2 Antibody (PCRP-LDB2-1B10), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24029AF647
This item is not returnable.
View return policy
Description
LDB2 Monoclonal antibody specifically detects LDB2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LDB2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Carboxyl-terminal LIM domain-binding protein 1, CLIM-1, CLIM1LDB1, LDB-2, LIM binding domain 2, LIM domain binding 2, LIM domain-binding factor CLIM1, LIM domain-binding factor-2, LIM domain-binding protein 2 | |
| Recombinant full-length human LDB2 protein | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9079 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-LDB2-1B10 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction