missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LDB2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567002

Product Code. 15989674

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SSTSNSSAGNN ANSTGSKKKT TAANLSLSSQ VPD Highest antigen sequence identity to the following orthologs - mouse 79%, rat 76%.

Genes encoding LIM domain-binding factors were initially isolated in a screen for proteins that physically interact with the LIM domains of nuclear proteins. These proteins, such as the one encoded by the LDB2 gene, are capable of binding to a variety of transcription factors and are likely to function at enhancers to bring together diverse transcription factors and form higher order activation complexes or to block formation of such complexes. The family of genes encoding LIM domain-binding factors includes 2 members isolated from the mouse, Clim1 and Clim2/Lbd1/Nli; and their homologs cloned from the frog, chicken, and fly. The fact that LIM domain-binding factors are likely to be involved in the coordination of the transcriptional activity of many diverse factors might implicate them in human phenotypes characterized by multiple affected sites.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LDB2
Polyclonal
Unconjugated
LDB2
AI035351; AW146358; Carboxyl-terminal LIM domain-binding protein 1; cLdb2; Clim1; CLIM-1; CLP-36; LDB1; Ldb2; LDB-2; Ldb3; LIM binding domain 2; LIM domain binding 2; LIM domain binding protein CLIM-1; LIM domain-binding factor CLIM1; LIM domain-binding factor-2; LIM domain-binding protein 2
Rabbit
Antigen affinity chromatography
RUO
9079
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O43679
LDB2
Recombinant Human LDB2. Recombinant protein control fragment (Product #RP-105455).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.