missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDB3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-2445
This item is not returnable.
View return policy
Description
LDB3 Polyclonal specifically detects LDB3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| LDB3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CMD1C, CYPHER, KIAA01613, KIAA0613FLJ35865, LDB3Z1, LDB3Z4, LIM domain binding 3, LIM domain-binding protein 3, ORACLE, PDLIM6, PDZ and LIM domain 6, Protein cypher, ZASPLVNC3, Z-band alternatively spliced PDZ-motif protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11155 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 1 ug/ml, Peptide ELISA Detection limit 1:1000 | |
| NP_009009.1 | |
| LDB3 | |
| Peptide with sequence C-KSQNKPEDEADE corresponding to internal region according to NP_009009.1, NP_001073583.1, NP_001073584.1, NP_001073585.1. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog and Pig . | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction