missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38431-100ul
This item is not returnable.
View return policy
Description
LDB3 Polyclonal antibody specifically detects LDB3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| LDB3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CMD1C, CYPHER, KIAA01613, KIAA0613FLJ35865, LDB3Z1, LDB3Z4, LIM domain binding 3, LIM domain-binding protein 3, ORACLE, PDLIM6, PDZ and LIM domain 6, Protein cypher, ZASPLVNC3, Z-band alternatively spliced PDZ-motif protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human LDB3 (NP_001073585.1).,, Sequence:, MSYSVTLTGPGPWGFRLQGGKDFNMPLTISRITPGSKAAQSQLSQGDLVVAIDGVNTDTMTHLEAQNKIKSASYNLSLTLQKSKRPIPISTTAPPVQTPLPVIPHQKVVVNSPANADYQERFNPSALKDSALSTHKPIEVKGLGGKATIIHAQYNTPISMYSQDAIMDAIAGQAQAQGSDFSGSLPIKDLAVDSASPVYQAVIKSQNKPEDEADEWARRSSNLQSRSFRILAQMTGTEFMQDPDEEALRRSRERFETERNSPRFAKLRNWHHGLSAQILNVKS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 11155 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction