Learn More
LDHA Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-21799-100UL
 View more versions of this product
Product Code. 18024245
  • € 450.00 / 100µL
Description
The LDHA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LDHA. This antibody recognizes human, mouse, rat antigen. The LDHA antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
LDHA
1000 μg/mL
lactate dehydrogenase A, LDH A, LDH M, LDH muscle subunit, LDH1, LDHA, PIG19
100 μL
3939
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa