missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LDHAL6A Polyclonal Antibody
GREENER_CHOICE

Product Code. 16607154
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607154 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16607154 Supplier Invitrogen™ Supplier No. PA5100667

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total LDHAL6A.

Displays an lactate dehydrogenase activity. Significantly increases the transcriptional activity of JUN, when overexpressed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LDHAL6A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LDHAL6A
Gene Accession No. Q6ZMR3
Gene Alias lactate dehydrogenase A like 6A; lactate dehydrogenase A-like 6A; LDH6A; LDHAL6A; LDHL2; L-lactate dehydrogenase A-like 6A
Gene Symbols LDHAL6A
Host Species Rabbit
Immunogen A synthesized peptide derived from human LDHAL6A(Accession Q6ZMR3), corresponding to amino acid residues L214-L264.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160287
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.