missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDHAL6B Antibody (1C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00092483-M04
This item is not returnable.
View return policy
Description
LDHAL6B Monoclonal antibody specifically detects LDHAL6B in Human samples. It is validated for Western Blot, ELISA
Specifications
| LDHAL6B | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 1.1.1, lactate dehydrogenase A-like 6, lactate dehydrogenase A-like 6B, LDH6B, LDHAL6, LDHLEC 1.1.1.27, L-lactate dehydrogenase A-like 6B | |
| LDHAL6B (NP_005497, 282 a.a. ∽ 381 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EVTATAYEIIKMKGYTSWAIGLSVADLTESILKNLRRIHPVSTITKGLYGIDEEVFLSIPCILGENGITNLIKIKLTPEEEAHLKKSAKTLWEIQNKLKL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1C9 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| NP_149972.1 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 92483 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction