Learn More
Invitrogen™ LDHB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA579594
Beschreibung
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human placenta tissue, human PC-3 whole cell, monkey COS-7 whole cell, human HEK293 whole cell, human A549 whole cell, monkey kidney tissue, rat kidney tissue, rat heart tissue, mouse kidney tissue, mouse heart tissue. IHC: mouse brain tissue, rat spleen tissue, human tonsil tissue. ICC/IF: A431 cell. Flow: A549 cell.
This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Spezifikation
| LDHB | |
| Polyclonal | |
| Unconjugated | |
| LDHB | |
| AI790582; epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; HEL-S-281; H-Ldh; lactate dehydrogenase B; lactate dehydrogenase H chain; lactate dehydrogenase-B; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; Ldh2; Ldh-2; LDHB; LDH-B; LDHBD; LDHH; LDH-H; L-lactate dehydrogenase B chain; Renal carcinoma antigen NY-REN-46; similar to lactate dehydrogenase B; testicular secretory protein Li 25; TRG-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16832, 24534, 3945 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P07195, P16125, P42123 | |
| LDHB | |
| E. coli-derived human LDHB recombinant protein (Position: E237-L334). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.