missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LDHB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582656
This item is not returnable.
View return policy
Description
Immunogen sequence: HPVSTMVKGM YGIENEVFLS LPCILNARGL TSVINQKLKD DEVAQLKKSA DTLWDIQKDL KDL Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
This gene encodes an enzyme which catalyzes the reversible conversion of lactate and pyruvate, and NAD and NADH, in the glycolytic pathway. Mutations in this gene are associated with lactate dehydrogenase B deficiency. Pseudogenes have been identified on the X chromosome and on chromosome. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| LDHB | |
| Polyclonal | |
| Unconjugated | |
| LDHB | |
| AI790582; epididymis secretory protein Li 281; H(B)-type lactate dehydrogenase; HEL-S-281; H-Ldh; lactate dehydrogenase B; lactate dehydrogenase H chain; lactate dehydrogenase-B; lactate dehydrogenase-H; lactate dehydrogenase-H (, EC 1.1.1.27); Lactate dehydrogenease B; LDH heart subunit; Ldh2; Ldh-2; LDHB; LDH-B; LDHBD; LDHH; LDH-H; L-lactate dehydrogenase B chain; Renal carcinoma antigen NY-REN-46; similar to lactate dehydrogenase B; testicular secretory protein Li 25; TRG-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07195 | |
| LDHB | |
| Recombinant protein corresponding to Human LDHB. Recombinant protein control fragment (Product #RP-102543). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction