missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLR Antibody (263123), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2255L
This item is not returnable.
View return policy
Description
LDLR Monoclonal antibody specifically detects LDLR in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| LDLR | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 263123 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LDL R, Ala22-Arg790 (Ala23Val, Cys27Gly), Accession # Q6GTJ9 | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
| 3949 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction