Learn More
Memmert™ Entry Port 38 mm for IF30plus
Product Code. 17536955 Shop All Memmert Products
Change view
Click to view available options
Diameter (Metric):
38 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
17536955 38 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17536955 Supplier Memmert™ Supplier No. IF30plusF7
Optional entry port for Memmert Incubators.
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
For Use With (Equipment) IF30plus Incubator
Type Entry Port
Diameter (Metric) 38 mm