Learn More
LDLRAP1 Polyclonal Antibody
GREENER_CHOICE
LDLRAP1 Polyclonal Antibody
Brand:  Invitrogen PA19008
Product Code. 11503851
  • € 530.00 / 100µg
Description
LDLRAP1 Polyclonal Antibody for ICC/IF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LDLRAP1
Polyclonal
Unconjugated
LDLRAP1
AA691260; Arh; Arh1; ARH2; Autosomal recessive hypercholesterolemia protein; autosomal recessive hypercholesterolemia protein homolog; DKFZp586D0624; FHCB1; FHCB2; LDL receptor adaptor protein; LDLRAP1; Low density lipoprotein receptor adapter protein 1; low density lipoprotein receptor adaptor protein 1; MGC34705; RGD1563417; RP11-70P17.2
Goat
Ammonium sulfate precipitation
RUO
26119
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q5SW96
LDLRAP1
Synthetic peptide corresponding to the N terminus amino acids DALKSAGRALIRS (aa 2-15).