missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDOC1 Antibody (OTI1A2), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71763G
This item is not returnable.
View return policy
Description
LDOC1 Monoclonal specifically detects LDOC1 in Human samples. It is validated for Western Blot.
Specifications
| LDOC1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LDOC1 | |
| Full length recombinant protein of human LDOC1 (NP_036449) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI1A2 | |
| Western Blot | |
| BCUR1, breast cancer, up-regulated 1, Leucine zipper protein down-regulated in cancer cells, leucine zipper, down-regulated in cancer 1, Mar7, Mart7, protein LDOC1 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 23641.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction