missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDOC1 Antibody (OTI1A2), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71763PE
This item is not returnable.
View return policy
Description
LDOC1 Monoclonal antibody specifically detects LDOC1 in Human samples. It is validated for Western Blot
Specifications
| LDOC1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI1A2 | |
| Western Blot | |
| BCUR1, breast cancer, up-regulated 1, Leucine zipper protein down-regulated in cancer cells, leucine zipper, down-regulated in cancer 1, Mar7, Mart7, protein LDOC1 | |
| Full length human recombinant protein of human LDOC1(NP_036449) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23641 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction