missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDOC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562475
This item is not returnable.
View return policy
Description
Immunogen sequence: NSQLMEQLRL LVCERASLLR QVRPPSCPVP FPETFNGESS RLPEFIV Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 74%.
LDOC1 Polyclonal antibody specifically detects LDOC1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| LDOC1 | |
| Polyclonal | |
| Unconjugated | |
| LDOC1 | |
| 7-Mar; BCUR1; breast cancer, up-regulated 1; Gm366; Ldoc1; LDOC1 regulator of NFKB signaling; LDOC1, regulator of NFKB signaling; Leucine zipper; leucine zipper downregulated in cancer; leucine zipper down-regulated in cancer 1; leucine zipper protein down-regulated in cancer cells; leucine zipper, down-regulated in cancer 1; Mar7; Mart7; Protein LDOC1; RGD1565644 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23641 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95751 | |
| LDOC1 | |
| Recombinant protein corresponding to Human LDOC1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction