Learn More
Buerkle™ LDPE Dropping Bottles
Product Code. 10267512 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. 10267512
Brand: Buerkle™ 03060030
Only null left
Add to basket
Add to basket
Dispense small amounts of liquid, drop by drop or as a fine jet. Buerkle™ LDPE Dropping Bottles feature a sturdy construction and convenient attached captive cap.
  • Manufactured from low density polyethylene (LDPE) for clarity, strength, and flexibility
  • Attached captive cap on polyethylene dropper cap fits tip and eliminates misplacement of cap
  • Dropper tip must be cut before first use for precision dropping
  • Suitable for use with foodstuff
  • Translucent
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 30 mL
Color Translucent
Diameter (Metric) Inner Neck 9 mm
For Use With (Application) Suitable for Use with Foodstuffs
Material LDPE
Thread Size (Metric) 14 mm
Closure Included Yes
Height (Metric) 96 mm
Closure Material Polyethylene
Diameter (Metric) 35 mm
Closure Type Captive Cap and Dropper Cap