Learn More
VITLAB™ Transparent LDPE Narrow-Mouth Bottles with Flat Shoulders
Product Code. 10054683
Click to view available options
Capacity (Metric):
10 mL
20 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
GL 14
GL 18
GL 25
GL 28
Unit Size:
Pack of 100
Pack of 25
Pack of 50
Product Code. 10054683
Brand: VITLAB™ 138493
Only null left
Add to basket
Add to basket
VITLAB™ Transparent LDPE Narrow-Mouth Bottles with Flat Shoulders are supplied with a screw cap. Made of LDPE, they provide a flexible, resilient storage option.
  • Product Type: Storage Bottle
  • Description: Narrow Mouth with Flat Shoulders
  • Material: LDPE
  • Shape: Round
  • Includes: Screw Cap
Specifications
Capacity (Metric) 100 mL
Color Transparent
Diameter (English) 1.8 in.
Material LDPE
Sterility Non-sterile
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 106 mm
Product Type Storage Bottle
Height (English) 4.2 in.
Closure Material LDPE
Closure Size GL 18
Diameter (Metric) 45 mm
Closure Type Screw Cap