Learn More
BRAND™ LDPE Screw Caps for Wide Mouth Bottles
10.50 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Use promo code "23264" to get your promotional price.
Click to view available options
Thread Size (Metric):
GL 32
GL 40
GL 50
For Use With (Equipment):
Fits GL 40 Thread wide mouth laboratory bottles
Description
- Manufactured from Low-Density Polyethylene (LDPE)
- Ridged for easy gripping
- Natural color
- Available in a variety of thread sizes
- For use with LDPE wide mouth laboratory bottles
Specifications
Material | LDPE |
Quantity | 25Pack |
Type | Screw Cap |
Thread Size (Metric) | GL 32 |
Certified | Yes |
For Use With (Equipment) | Fits GL 32 Thread wide mouth laboratory bottles |