Learn More
Poulten & Graf GmbH LDPE-Stopper
Poulten & Graf GmbH LDPE-Stopper uses a sealing profile to secure the neck of a flask or bottle.
Brand: Poulten & Graf GmbH 10144445
Additional Details : Weight : 0.10000kg
Description
- Low-density polyethylene
- With sealing profile
- According to DIN 12254
Specifications
DIN 12254 | |
Flasks and bottles | |
TS 45/40 | |
Polyethylene |
PP-Stopper | |
10 | |
TS 45/40 |