Learn More
Poulten & Graf GmbH LDPE-Stopper
Click to view available options
Size:
TS 34/35
TS 45/40
TS 60/46
Unit Size:
Each
Description
- Low-density polyethylene
- With sealing profile
- According to DIN 12254
Specifications
Specifications
| Certifications/Compliance | DIN 12254 |
| Description | PP-Stopper |
| For Use With (Equipment) | Flasks and bottles |
| Quantity | 10 |
| Size | TS 45/40 |
| Standard Taper Joint | TS 45/40 |
| Material | Polyethylene |